Abnova
Manufacturer Code:PAB18556
Catalog # 89-120-113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 400-410 at internal region of human GABRG2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of GABRG2.
For Research Use Only
Protein Aliases: CAE2; ECA2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2; GEFSP3
Gene Aliases: CAE2; ECA2; GABRG2; GEFSP3
UniProt ID: (Human) P18507
Entrez Gene ID: (Human) 2566
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.