Abnova
Manufacturer Code:H00002564M01A
Catalog # 89-104-525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G11 |
Immunogen | GABRE (AAH59376.11 a.a. &bsim 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GABRE.
For Research Use Only
Protein Aliases: GABA(A) receptor subunit epsilon; GABA(A) receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; Gamma-aminobutyric acid receptor subunit epsilon
Gene Aliases: GABRE
UniProt ID: (Human) P78334
Entrez Gene ID: (Human) 2564
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.