Invitrogen
Manufacturer Code:PA530255
Catalog # PIPA530255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 173 of Human GABA Receptor Epsilon |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit epsilon; GABA(A) receptor subunit epsilon GABA(A) receptor epsilon gamma-aminobutyric acid receptor subunit epsilon; GABA(A) receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; Gamma-aminobutyric acid receptor subunit epsilon
Gene Aliases: GABRE
UniProt ID: (Human) P78334
Entrez Gene ID: (Human) 2564
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.