Proteintech
Manufacturer Code:201831AP150UL
Catalog # 50-173-2754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GABRB1 Fusion Protein Ag14070 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA A receptor beta 1 GABA A receptor beta GABAA receptor beta 1 GABRB1; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; Gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1
Gene Aliases: AW061132; B230208N19Rik; Gabrb-1; GABRB1; GARB1
UniProt ID: (Human) P18505, (Mouse) P50571, (Rat) P15431
Entrez Gene ID: (Human) 2560, (Mouse) 14400, (Rat) 25450
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.