Invitrogen
Manufacturer Code:PA5106667
Catalog # PIPA5106667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GABRA6 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha6 GABA subunit A receptor alpha 6 GABA(A) receptor subunit alpha-6 GABA(A) receptor alpha 6 GABA-A receptor alpha-6 subunit GABAAR alpha6 GABA-ARalpha6 GABRA 6 Gabra6 Gabra-6 gamma-aminobutyric acid gamma-aminobutyric acid A receptor alpha 6 gamma-aminobutyric acid (GABA) A receptor alpha 6 gamma-aminobutyric acid (GABA) A receptor subunit alpha 6 gamma-aminobutyric acid (GABA-A) receptor subunit alpha 6 gamma-aminobutyric acid A receptor alpha 6 gamma-aminobutyric acid receptor subunit alpha-6 gamma-aminobutyric acid type A receptor alpha6 subunit subunit alpha 6 gamma-aminobutyric acid (GABA-A) receptor; GABA subunit A receptor alpha 6; GABA(A) receptor subunit alpha-6; GABA(A) receptor, alpha 6; GABA-A receptor alpha-6 subunit; gamma-aminobutyric acid A receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, alpha 6; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 6; gamma-aminobutyric acid A receptor, alpha 6; Gamma-aminobutyric acid receptor subunit alpha-6; subunit alpha 6 gamma-aminobutyric acid (GABA-A) receptor
Gene Aliases: alpha6; GABA-ARalpha6; Gabra-6; GABRA6
UniProt ID: (Human) Q16445, (Mouse) P16305, (Rat) P30191
Entrez Gene ID: (Human) 2559, (Mouse) 14399, (Rat) 29708
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.