Invitrogen
Manufacturer Code:PA577407
Catalog # PIPA577407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide NSKDEKLSPENFTR(C) corresponding to amino acid residues 37-50 of rat GABA(A) alpha4 with replacement of C44 with serine |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-4; GABA(A) receptor, alpha 4; GABA-A receptor alpha-4 subunit; GABRA4; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 4; gamma-aminobutyric acid A receptor alpha 4; Gamma-aminobutyric acid receptor subunit alpha-4
Gene Aliases: Gabra-4; GABRA4
UniProt ID: (Human) P48169, (Mouse) Q9D6F4, (Rat) P28471
Entrez Gene ID: (Human) 2557, (Mouse) 14397, (Rat) 140675
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.