Invitrogen
Manufacturer Code:PA540998
Catalog # PIPA540998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GABRA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABA-A receptor alpha-3 subunit; Gamma-aminobutyric acid (GABA) A receptor alpha 3; gamma-aminobutyric acid (GABA) A receptor, alpha 3; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 3; Gamma-aminobutyric acid receptor subunit alpha-3; gamma-aminobutyric acid receptor, subunit alpha 3; gamma-aminobutyric acid type A receptor alpha3 subunit; Gamma-animobutyric acid (GABA) A receptor alpha 3; Gamma-animobutyric acid (GABA) A receptor, alpha 3; unnamed protein product
Gene Aliases: Gabra-3; GABRA3
UniProt ID: (Bovine) P10064, (Human) P34903, (Mouse) P26049, (Rat) P20236
Entrez Gene ID: (Bovine) 282237, (Dog) 492223, (Horse) 100063420, (Rabbit) 100349107, (Human) 2556, (Mouse) 14396, (Rat) 24947, (Guinea pig) 100725783
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.