Invitrogen
Manufacturer Code:PA533568
Catalog # PIPA533568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from C-Terminus of human GABRA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; GABRA3 GABA(A) receptor alpha 3 GABA A receptor alpha 3; gamma-aminobutyric acid (GABA) A receptor, alpha 3; Gamma-aminobutyric acid receptor subunit alpha-3; gamma-animobutyric acid (GABA) A receptor, alpha 3
Gene Aliases: GABRA3
UniProt ID: (Human) P34903
Entrez Gene ID: (Human) 2556
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.