Invitrogen
Manufacturer Code:PA526305
Catalog # PIPA526305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 378-406 amino acids from the C-terminal region of human GABRA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-2; GABA(A) receptor subunit alpha-2 GABA(A) receptor alpha 2 gamma-aminobutyric acid receptor subunit alpha-2; GABA(A) receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid A receptor alpha 2; Gamma-aminobutyric acid receptor subunit alpha-2
Gene Aliases: GABRA2
UniProt ID: (Human) P47869
Entrez Gene ID: (Human) 2555
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.