Invitrogen
Manufacturer Code:PA5109750
Catalog # PIPA5109750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABRA2 (NP_000798.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C630048P16Rik FLJ97076 GABA(A) receptor GABA(A) receptor subunit alpha-2 GABAARalpha2 GABRA2 Gabra-2 Gamma-aminobutyric acid receptor subunit alpha-2; GABA(A) receptor subunit alpha-2; GABA(A) receptor, alpha 2; gamma-aminobutyric acid (GABA) A receptor, alpha 2; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 2; gamma-aminobutyric acid A receptor alpha 2; gamma-aminobutyric acid receptor A2 subunit; Gamma-aminobutyric acid receptor subunit alpha-2
Gene Aliases: C630048P16Rik; Gabra-2; GABRA2
UniProt ID: (Human) P47869, (Mouse) P26048
Entrez Gene ID: (Human) 2555, (Mouse) 14395, (Rat) 289606
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.