Invitrogen
Manufacturer Code:PA5114315
Catalog # PIPA5114315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within rat GABRA1 aa 1-50. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECA4 EIEE19 EJM EJM5 GABA(A) receptor subunit alpha-1 GABA(A) receptor alpha 1 GABAA alpha 1 GABAAR alpha1 GABRA 1 GABRA1 Gabra-1 gamma-aminobutyric acid gamma-aminobutyric acid A receptor alpha 1 gamma-aminobutyric acid (GABA) A receptor alpha 1 gamma-aminobutyric acid (GABA) A receptor subunit alpha 1 gamma-aminobutyric acid (GABA-A) receptor subunit alpha 1 gamma-aminobutyric acid A receptor alpha 1 Gamma-aminobutyric acid receptor subunit alpha-1 gamma-aminobutyric acid type A receptor alpha1 subunit; GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor, alpha 1; Gamma-aminobutyric acid receptor subunit alpha-1
Gene Aliases: ECA4; EIEE19; EJM; EJM5; Gabra-1; GABRA1
UniProt ID: (Human) P14867, (Rat) P62813
Entrez Gene ID: (Human) 2554, (Rat) 29705
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.