Invitrogen
Manufacturer Code:PA523720
Catalog # PIPA523720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 861-890 amino acids from the C-terminal region of human GABBR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 51; G-protein coupled receptor 51; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABA-B-R2; GABA-BR2; GABABR2 GPR51 GPRC3B HG20 HRIHFB2099 G protein-coupled receptor 51 G-protein coupled receptor 51 GABA-B receptor 2 GABA-B receptor R2 subunit GABA-B-R2 GABA-BR2 gamma-aminobutyric acid type B receptor subunit 2 gb2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; Gamma-aminobutyric acid type B receptor subunit 2; gamma-aminobutyric acid type B receptor, subunit 2; gb2; HG20; ortholog of human G protein-coupled receptor 51 GPR51
Gene Aliases: GABABR2; GABBR2; Gb2; Gm425; GPR51; GPRC3B; HG20; HRIHFB2099
UniProt ID: (Human) O75899, (Mouse) Q80T41
Entrez Gene ID: (Human) 9568, (Mouse) 242425
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.