Invitrogen
Manufacturer Code:PA533566
Catalog # PIPA533566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GABBR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA-B receptor 1; GABA-B receptor, R1 subunit; GABBR1 DJ271M21.1.1 DJ271M21.1.2 GABA(B) receptor Gaba-b-r1a GABA-BR1 Gaba-br1a receptor GABAB subunit 1c GABAB1 GABA-B receptor 1 Gaba-b1 Gababr1 receptor GPRC3A Gabab-receptor subunit 1a GABABR1 GABBR1-3 GBR1 HGB1a GABA-B-R1 GB1; gamma-aminobutyric acid (GABA) B receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; seven transmembrane helix receptor
Gene Aliases: GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A
UniProt ID: (Human) O95468
Entrez Gene ID: (Human) 2550
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.