Invitrogen
Manufacturer Code:PA132248
Catalog # PIPA132248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | N-terminus of GABA BR1 subunit (Proprietary) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAAP-188P13.3 FLJ92613 GABABR1 GABBR1-3 GB1 GPRC3A dJ271M21.1.1 dJ271M21.1.2; GABA-B receptor 1; GABA-B receptor, R1 subunit; gamma-aminobutyric acid (GABA) B receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; seven transmembrane helix receptor
Gene Aliases: GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A
UniProt ID: (Human) O95468
Entrez Gene ID: (Human) 2550
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.