Invitrogen
Manufacturer Code:PA110031
Catalog # PIPA110031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (coupled with THG) derived from amino acid sequence 30-47 of mouse GABA B receptor 1b. Only 1 amino acid is different from immunogen derived from human GB1b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); GABA-B receptor 1; GABA-B-R1; GABA-BR1; GABA-BR1b receptor; GABAB1 GABAbR1 bM573K1.1 GABA-B receptor 1 GABA-B-R1 GABA-BR1 GABA-BR1b receptor bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor 1a) gamma-aminobutyric acid (GABA-B) receptor 1 gamma-aminobutyric acid type B receptor subunit 1 gb1; GABABR1; gamma-aminobutyric acid (GABA-B) receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; gb1
Gene Aliases: bM573K1.1; GABAB1; GABAbR1; Gabbr1
UniProt ID: (Mouse) Q9WV18, (Rat) Q9Z0U4
Entrez Gene ID: (Mouse) 54393, (Rat) 81657
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.