Invitrogen
Manufacturer Code:MA535814
Catalog # PIMA535814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1648 |
Immunogen | A synthesized peptide derived from human GABARAPL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 0610012F20Rik 2900019O08Rik AI173605 ATG8 ATG8C FLC3A GABA(A) receptor-associated protein-like 2 Gabarapl2 Gamma-aminobutyric acid receptor-associated protein-like 2 Ganglioside expression factor 2 GATE16 GATE-16 Gef2 GEF-2 Genera General protein transport factor p16 Golgi-associated ATPase enhancer of 16 kDa MAP1 light chain 3-related protein zgc:92319; GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; gamma-aminobutyric acid (GABA-A) receptor-associated protein-like 2; Gamma-aminobutyric acid receptor-associated protein-like 2; Ganglioside expression factor 2; GATE-16; GEF-2; General protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein
Gene Aliases: 0610012F20Rik; 2900019O08Rik; AI173605; ATG8; ATG8C; FLC3A; GABARAPL2; GATE-16; GATE16; GEF-2; GEF2
UniProt ID: (Human) P60520, (Mouse) P60521, (Rat) P60522
Entrez Gene ID: (Human) 11345, (Mouse) 93739, (Rat) 64670
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.