Invitrogen
Manufacturer Code:PA580650
Catalog # PIPA580650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GATE-16/GABARAPL2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor-associated protein like 2; GABA(A) receptor-associated protein-like 2; Gamma-aminobutyric acid receptor-associated protein-like 2; Gamma-aminobutyric acid receptor-associated protein-like 2 GABA(A) receptor-associated protein-like 2 Ganglioside expression factor 2 GEF-2 Genera; Ganglioside expression factor 2; GATE-16; GEF-2; General protein transport factor p16; Golgi-associated ATPase enhancer of 16 kDa; MAP1 light chain 3 related protein; MAP1 light chain 3-related protein
Gene Aliases: ATG8; ATG8C; FLC3A; GABARAPL2; GATE-16; GATE16; GEF-2; GEF2
UniProt ID: (Human) P60520
Entrez Gene ID: (Human) 11345
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.