Novus Biologicals
Manufacturer Code:NBP304414100UL
Catalog # NB094291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human GABARAPL1 (NP_113600.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG8L APG8-LIKE ATG8 ATG8B ATG8L Early estrogen-regulated protein GABA(A) receptor-associated protein like 1 gamma-aminobutyric acid receptor-associated protein-like 1 gec1 GEC-1 GEC1GABA(A) receptor-associated protein-like 1 Glandular epithelial cell protein 1; Early estrogen-regulated protein; GABA(A) receptor-associated protein like 1; GABA(A) receptor-associated protein-like 1; GABARAPL1-a; Gamma-aminobutyric acid receptor-associated protein-like 1; GEC-1; Glandular epithelial cell protein 1
Gene Aliases: APG8-LIKE; APG8L; ATG8; ATG8B; ATG8L; GABARAPL1; GEC1
UniProt ID: (Human) Q9H0R8
Entrez Gene ID: (Human) 23710
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.