Novus Biologicals
Manufacturer Code:NBP266854
Catalog # NBP266854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM30-30 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ATG8A FLC3B FLJ25768 GABA(A) receptor-associated proteinMGC120154 gamma-aminobutyric acid receptor-associated protein MM46MGC120155; GABA(A) receptor-associated protein; Gamma-aminobutyric acid receptor-associated protein; hypothetical protein; MM46
Gene Aliases: ATG8A; FLC3B; GABARAP; GABARAP-a; HT004; MM46
UniProt ID: (Human) O95166
Entrez Gene ID: (Human) 11337
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.