Invitrogen
Manufacturer Code:OSG00005W
Catalog # PIOSG00005W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human GABARAP conjugated to an immunogenic carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor-associated protein; GABA(A) receptor-associated protein MM46 GABARAP FLC3B HT004 FLJ25768 MGC120154 MGC120155; GABA-A receptor-associated protein; gamma-aminobutyric acid receptor associated protein; Gamma-aminobutyric acid receptor-associated protein; gamma-aminobutyric acid reseptor associated protein; hypothetical protein; MM46
Gene Aliases: ATG8A; FLC3B; GABARAP; GABARAP-a; HT004; MM46
UniProt ID: (Human) O95166, (Rat) P60517, (Mouse) Q9DCD6
Entrez Gene ID: (Human) 11337, (Rat) 58974, (Mouse) 56486
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.