Abnova
Manufacturer Code:PAB27437
Catalog # 89-128-947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at N-terminus of human GABARAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GABARAP.
For Research Use Only
Protein Aliases: FLJ25768; GABA(A) receptor-associated protein; Gamma-aminobutyric acid receptor-associated protein; hypothetical protein; MGC120154; MGC120155; MM46
Gene Aliases: ATG8A; FLC3B; GABARAP; GABARAP-a; HT004; MM46
UniProt ID: (Human) O95166
Entrez Gene ID: (Human) 11337
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.