Novus Biologicals
Manufacturer Code:NBP259323C
Catalog # NBP259323C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S413-67 |
Immunogen | Fusion protein amino acids 27-253 (Extracellular N-terminus) of human GABA A Receptor Epsilon |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GABA(A) receptor subunit epsilon; GABA(A) receptor subunit epsilon gamma-aminobutyric acid (GABA) A receptor epsilon gamma-aminobutyric acid receptor subunit epsilon; GABA(A) receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; Gamma-aminobutyric acid receptor subunit epsilon
Gene Aliases: GABRE
UniProt ID: (Human) P78334
Entrez Gene ID: (Human) 2564
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.