Novus Biologicals
Manufacturer Code:NBP259335
Catalog # NBP259335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S81-2 |
Immunogen | Fusion protein amino acids 861-912 of rat GABA(B)R2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 51; G protein-coupled receptor 51 GABA-BR2 GABA-B-R2 GABABR2GABA-B receptor 2 gamma-aminobutyric acid (GABA) B receptor 2 gamma-aminobutyric acid type B receptor subunit 2 Gb2 GPR51GABA-B receptor R2 subunit GPRC3BFLJ36928 G-protein coupled receptor 51 HG20HRIHFB2099; G-protein coupled receptor 51; GABA-B receptor 2; GABA-B receptor, R2 subunit; GABA-B-R2; GABA-BR2; gamma-aminobutyric acid (GABA) B receptor, 2; gamma-aminobutyric acid B receptor 2; Gamma-aminobutyric acid type B receptor subunit 2; gb2; HG20
Gene Aliases: GABABR2; GABBR2; GPR51; GPRC3B; HG20; HRIHFB2099
UniProt ID: (Human) O75899
Entrez Gene ID: (Human) 9568
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.