Novus Biologicals
Manufacturer Code:NBP259336AF700
Catalog # NB259336700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S93A-49 |
Immunogen | Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ92613 GABABR1 GABBR1-3 gamma-aminobutyric acid (GABA) B receptor 1 gamma-aminobutyric acid type B receptor subunit 1 GB1 GPRC3Asubunit 1c hGB1a seven transmembrane helix receptor; GABA-B receptor 1; GABA-B receptor, R1 subunit; gamma-aminobutyric acid (GABA) B receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; seven transmembrane helix receptor
Gene Aliases: GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A
UniProt ID: (Human) O95468
Entrez Gene ID: (Human) 2550
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.