Novus Biologicals
Manufacturer Code:NB300196
Catalog # NB300196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein from the cytoplasmic loop of the alpha 6 subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA subunit A receptor alpha 6; GABA subunit A receptor alpha 6 GABA(A) receptor subunit alpha-6 gamma-aminobutyric acid (GABA) A receptor alpha 6 gamma-aminobutyric acid receptor subunit alpha-6 MGC116903 MGC116904; GABA(A) receptor subunit alpha-6; GABA(A) receptor, alpha 6; gamma-aminobutyric acid (GABA) A receptor, alpha 6; Gamma-aminobutyric acid receptor subunit alpha-6
Gene Aliases: GABRA6
UniProt ID: (Human) Q16445
Entrez Gene ID: (Human) 2559
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.