Novus Biologicals
Manufacturer Code:NBP232077
Catalog # NBP232077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 425 to 475 of human GABA A Receptor alpha 4 Antibody Alpha 4 using the numbering given in entry NP_000800.2 (GeneID 2557). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-4; GABA(A) receptor subunit alpha-4 gamma-aminobutyric acid (GABA) A receptor alpha 4 gamma-aminobutyric acid receptor subunit alpha-4; GABA(A) receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid A receptor alpha 4; Gamma-aminobutyric acid receptor subunit alpha-4
Gene Aliases: GABRA4
UniProt ID: (Human) P48169
Entrez Gene ID: (Human) 2557
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.