Novus Biologicals
Manufacturer Code:NB300193
Catalog # NB300193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein from the cytoplasmic loop of the alpha 4 subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GABA(A) receptor subunit alpha-4; GABA(A) receptor subunit alpha-4 gamma-aminobutyric acid (GABA) A receptor alpha 4 gamma-aminobutyric acid receptor subunit alpha-4; GABA(A) receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid A receptor alpha 4; Gamma-aminobutyric acid receptor subunit alpha-4
Gene Aliases: GABRA4
UniProt ID: (Human) P48169
Entrez Gene ID: (Human) 2557
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.