Novus Biologicals
Manufacturer Code:NB300151
Catalog # NB300151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acid residues specific to the gamma 2 subunit conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAE2 ECA2 GABA(A) receptor subunit gamma-2 gamma-aminobutyric acid (GABA) A receptor gamma 2 gamma-aminobutyric acid receptor subunit gamma-2 GEFSP3; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2
Gene Aliases: CAE2; ECA2; GABRG2; GEFSP3
UniProt ID: (Human) P18507
Entrez Gene ID: (Human) 2566
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.