Abnova
Manufacturer Code:PAB17534
Catalog # 89-119-195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human GAB4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GAB4.
For Research Use Only
Protein Aliases: GAB2-like; GRB2-associated binder 2-like; GRB2-associated binder 2-like protein; GRB2-associated binder 4; GRB2-associated binding protein family, member 4; GRB2-associated-binding protein 2-like; GRB2-associated-binding protein 4; Growth factor receptor bound protein 2-associated protein 4
Gene Aliases: GAB4
UniProt ID: (Human) Q2WGN9
Entrez Gene ID: (Human) 128954
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.