Invitrogen
Manufacturer Code:PA588958
Catalog # PIPA588958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 377-676 of human GAB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Grb2 associated binder 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; Growth factor receptor bound protein 2-associated protein 2; PH domain-containing adaptor molecule p97; pp100
Gene Aliases: AI463667; D130058I17Rik; GAB2; KIAA0571; p97
UniProt ID: (Human) Q9UQC2
Entrez Gene ID: (Human) 9846, (Mouse) 14389
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.