Novus Biologicals
Manufacturer Code:NBP255547
Catalog # NBP255547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSSSSQHLLRERKSSAPSHSSQPTLFTFEPPVSNHMQPTLSTSAPQEYLYLHQCISRRAENARSASFSQGTRASFLMRSDTAVQKLAQGNGHCVNGISGQVHGFYSLPKPSRHNTEFRDST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated binding protein 2 GRB2-associated-binding protein 2 Growth factor receptor bound protein 2-associated protein 2 KIAA0571GRB2-associated binder 2 pp100; GRB2-associated-binding protein 2; Growth factor receptor bound protein 2-associated protein 2; pp100
Gene Aliases: GAB2; KIAA0571
UniProt ID: (Human) Q9UQC2
Entrez Gene ID: (Human) 9846
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.