Novus Biologicals
Manufacturer Code:NBP304453100UL
Catalog # NB095945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GAB1 (NP_997006.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GRB2-associated binder 1; GRB2-associated binder 1 GRB2-associated binding protein 1 GRB2-associated-binding protein 1 Growth factor receptor bound protein 2-associated protein 1; GRB2-associated binding protein 1; GRB2-associated-binding protein 1; Growth factor receptor bound protein 2-associated protein 1
Gene Aliases: GAB1
UniProt ID: (Human) Q13480
Entrez Gene ID: (Human) 2549
Molecular Function: transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.