Invitrogen
Manufacturer Code:MA537870
Catalog # PIMA537870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2392 |
Immunogen | Recombinant protein of human GAA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa lysosomal alpha-glucosidase; 70 kDa lysosomal alpha-glucosidase 76 kDa lysosomal alpha-glucosidase acid (Pompe disease glycogen storage disease type II) acid alpha-glucosidase acid maltase Aglucosidase alfa E430018M07Rik Gaa glucosidase alpha acid glucosidase alpha acid glucosidase alpha acid (Pompe disease glycogen storage disease type II) LYAG lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; Acid maltase; Aglucosidase alfa; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); Lysosomal alpha-glucosidase
Gene Aliases: E430018M07Rik; GAA; LYAG
UniProt ID: (Human) P10253, (Mouse) P70699, (Rat) Q6P7A9
Entrez Gene ID: (Human) 2548, (Mouse) 14387, (Rat) 367562
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.