Invitrogen
Manufacturer Code:MA536228
Catalog # PIMA536228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G7 |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Synaptophysin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa lysosomal alpha-glucosidase; 70 kDa lysosomal alpha-glucosidase 76 kDa lysosomal alpha-glucosidase acid (Pompe disease glycogen storage disease type II) acid alpha-glucosidase acid maltase Aglucosidase alfa E430018M07Rik Gaa glucosidase alpha acid glucosidase alpha acid glucosidase alpha acid (Pompe disease glycogen storage disease type II) LYAG lysosomal alpha-glucosidase; 76 kDa lysosomal alpha-glucosidase; Acid maltase; Aglucosidase alfa; Lysosomal alpha-glucosidase
Gene Aliases: GAA; LYAG
UniProt ID: (Human) P10253
Entrez Gene ID: (Human) 2548
Molecular Function:
glucosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.