Novus Biologicals
Manufacturer Code:NBP242846
Catalog # NBP242846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of mouse G9a. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat-containing protein; ankyrin repeat-containing protein BAT8 C6orf30 chromosome 6 open reading frame 30 DKFZp686H08213 EC 2.1.1.- EC 2.1.1.43 euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8 FLJ35547 G9A histone methyltransferase G9AEm:AF134726.3 H3-K9-HMTase 3 Histone H3-K9 methyltransferase 3 histone-lysine N-methyltransferase H3 lysine-9 specific 3 HLA-B associated transcript 8 KMT1CNG36/G9a Lysine N-methyltransferase 1C NG36 Protein G9a; Euchromatic histone-lysine N-methyltransferase 2; G9A histone methyltransferase; H3-K9-HMTase 3; Histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B-associated transcript 8; Lysine N-methyltransferase 1C; Protein G9a
Gene Aliases: BAT8; C6orf30; EHMT2; G9A; GAT8; KMT1C; NG36
UniProt ID: (Human) Q96KQ7
Entrez Gene ID: (Human) 10919
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.