Invitrogen
Manufacturer Code:MA515921
Catalog # PIMA515921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H7 |
Immunogen | Purified recombit fragment of human G6PD expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G6PD; G6PD1 glucose-6-phosphate 1-dehydrogenase glucose-6-phosphate dehydrogenase G6PD; Glucose-6-phosphate 1-dehydrogenase
Gene Aliases: G6PD; G6PD1
UniProt ID: (Human) Q16000
Entrez Gene ID: (Human) 2539
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.