Invitrogen
Manufacturer Code:PA518734
Catalog # PIPA518734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (STNSDDVRDEKVK) corresponding to the internal amino acids of G6PD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: G6PD; G6PD glucose-6-phosphate dehydrogenase G6PD1 glucose-6-phosphate 1-dehydrogenase glucose-6-phosphate dehydrogenase G6PD; Glucose-6-phosphate 1-dehydrogenase; Glucose-6-phosphate 1-dehydrogenase X
Gene Aliases: G28A; G6PD; G6pd-1; G6PD1; G6pdx; Gpdx
UniProt ID: (Human) Q16000, (Mouse) Q00612
Entrez Gene ID: (Human) 2539, (Mouse) 14381
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.