Invitrogen
Manufacturer Code:PA570653
Catalog # PIPA570653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human G6PC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-6-Pase 3; G6Pase 3; G6Pase-beta; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; Glucose-6-phosphatase 3; Glucose-6-phosphatase beta; glucose-6-phosphatase catalytic subunit 3; glucose-6-phosphatase catalytic subunit-related; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein; UGRP
Gene Aliases: 0710001K01Rik; AU019276; AU045429; AV128920; AW545836; G6PC3; SCN4; UGRP; zgc:158425
UniProt ID: (Human) Q9BUM1, (Bovine) Q148G2, (Rat) Q6AZ83, (Mouse) Q6NSQ9, (Zebrafish) A1A5Z0
Entrez Gene ID: (Dog) 490942, (Human) 92579, (Horse) 100064862, (Rabbit) 100358721, (Sheep) 101113421, (Bovine) 369023, (Rat) 303565, (Guinea pig) 100723248, (Mouse) 68401, (Zebrafish) 790919
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.