Novus Biologicals
Manufacturer Code:NBP303956100UL
Catalog # NB094177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 50-150 of human G6PC3 (NP_612396.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.9 G6Pase 3 G-6-Pase 3 G6Pase-beta glucose 6 phosphatase catalytic 3 glucose-6-phosphatase 3 Glucose-6-phosphatase beta glucose-6-phosphatase catalytic subunit 3 Ubiquitous glucose-6-phosphatase catalytic subunit-related protein ubiquitously expressed G6Pase catalytic subunit-related protein UGRPSCN4; G-6-Pase 3; G6Pase 3; G6Pase-beta; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; Glucose-6-phosphatase 3; Glucose-6-phosphatase beta; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein
Gene Aliases: G6PC3; SCN4; UGRP
UniProt ID: (Human) Q9BUM1
Entrez Gene ID: (Human) 92579
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.