Abnova
Manufacturer Code:H00092579K
Catalog # 89-208-602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human G6PC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human G6PC3 peptide using ARM Technology.
For Research Use Only
Protein Aliases: G-6-Pase 3; G6Pase 3; G6Pase-beta; glucose 6 phosphatase catalytic subunit 3; glucose 6 phosphatase, catalytic, 3; Glucose-6-phosphatase 3; Glucose-6-phosphatase beta; Ubiquitous glucose-6-phosphatase catalytic subunit-related protein; ubiquitously expressed G6Pase catalytic subunit-related protein; UGRP
Gene Aliases: G6PC3; SCN4; UGRP
UniProt ID: (Human) Q9BUM1
Entrez Gene ID: (Human) 92579
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.