Invitrogen
Manufacturer Code:720253
Catalog # PI720253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptides corresponding to Human G6PC2 (aa 2-20 341-354) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-6-Pase 2; G6Pase 2; Glucose-6-phosphatase 2; Glucose-6-Phosphatase Catalytic Subunit 2; glucose-6-phosphatase, catalytic, 2; glucose-6-phosphatase, catalytic, related sequence; islet-specific G6CP-related protein; Islet-specific glucose-6-phosphatase catalytic subunit-related protein; islet-specific glucose-6-phosphatase-related protein; similar to glucose-6-phosphatase, catalytic, related
Gene Aliases: G6pc-rs; G6PC2; IGRP
UniProt ID: (Human) Q9NQR9, (Mouse) Q9Z186
Entrez Gene ID: (Human) 57818, (Mouse) 14378, (Rat) 681817
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.