Invitrogen
Manufacturer Code:PA542541
Catalog # PIPA542541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human G6PC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-6-Pase; G6Pase; G6Pase-alpha; Glc-6-Pase-alpha; Glucose-6-phosphatase; Glucose-6-phosphatase alpha; Glucose-6-phosphatase catalytic subunit 1; Glucose-6-phosphatase catalytic subunit G6PT GSD1a MGC163350 GSD1 G6PC1; glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierke disease); glucose-6-phosphatase, catalytic subunit
Gene Aliases: AW107337; G6Pase; G6PC; G6PC1; G6PT; Glc-6-Pase; GSD1; GSD1a
UniProt ID: (Human) P35575, (Bovine) Q29RU6, (Rat) P43428, (Mouse) P35576
Entrez Gene ID: (Horse) 100052233, (Human) 2538, (Rabbit) 100303766, (Sheep) 100037689, (Pig) 100134959, (Dog) 403492, (Bovine) 538710, (Rat) 25634, (Guinea pig) 100720420, (Mouse) 14377
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.