Invitrogen
Manufacturer Code:PA525148
Catalog # PIPA525148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 123-149 amino acids from the central region of human G6PC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-6-Pase; G6Pase-alpha; G6PC1 G6PT GSD1 GSD1a G-6-Pase G6Pase G6Pase-alpha glucose-6-phosphatase glucose-6-phosphatase alpha; Glucose-6-phosphatase; Glucose-6-phosphatase alpha; Glucose-6-phosphatase catalytic subunit 1; glucose-6-phosphatase, catalytic subunit
Gene Aliases: G6Pase; G6PC; G6PC1; G6PT; GSD1; GSD1a
UniProt ID: (Human) P35575
Entrez Gene ID: (Human) 2538
Molecular Function: carbohydrate phosphatase hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.