Proteintech
Manufacturer Code:221691AP150UL
Catalog # 50-173-3755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | G6PC Fusion Protein Ag17839 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G 6 Pase G6Pase G6Pase alpha G6PC G6PT Glucose 6 phosphatase Glucose 6 phosphatase alpha GSD1 GSD1a; G-6-Pase; G6Pase-alpha; Glucose-6-phosphatase; Glucose-6-phosphatase alpha; Glucose-6-phosphatase catalytic subunit 1; glucose-6-phosphatase, catalytic subunit
Gene Aliases: G6Pase; G6PC; G6PC1; G6PT; GSD1; GSD1a
UniProt ID: (Human) P35575
Entrez Gene ID: (Human) 2538
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.