Invitrogen
Manufacturer Code:PA539397
Catalog # PIPA539397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human G3BP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G3BP-2; GAP SH3 domain-binding protein 2; GTPase activating protein (SH3 domain) binding protein 2; KIAA0660; Ras GTPase-activating protein-binding protein 2; Ras-GTPase activating protein SH3 domain-binding protein 2
Gene Aliases: AA409541; E430034L04Rik; G3BP; G3BP2; KIAA0660; mKIAA0660; RGD1309571
UniProt ID: (Human) Q9UN86, (Mouse) P97379
Entrez Gene ID: (Human) 9908, (Rat) 305240, (Mouse) 23881
Molecular Function:
RNA binding protein
nucleic acid binding
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.