Novus Biologicals
Manufacturer Code:NBP15528120UL
Catalog # NB15739520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to KIAA1333(KIAA1333) The peptide sequence was selected from the N terminal of KIAA1333. Peptide sequence IWQRGKEEEGVYGFLIEDIRKEVNRASKLKCCVCKKNGASIGCVAPRCKR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.3.2 EC 6.3.2.- FLJ20333 G2/M phase-specific E3 ubiquitin-protein ligase G2/M-phase specific E3 ubiquitin ligase G2/M-phase specific E3 ubiquitin protein ligase KIAA1333PHD finger protein 7B PHF7B; G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; PHD finger protein 7B
Gene Aliases: G2E3; KIAA1333; PHF7B
UniProt ID: (Human) Q7L622
Entrez Gene ID: (Human) 55632
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.