Invitrogen
Manufacturer Code:PA5109748
Catalog # PIPA5109748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human G0S2 (NP_056529.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI255151 AV006465 G0/G1 switch protein 2 G0/G1 switch regulatory protein 2 G0S2 G0S2-like protein Putative lymphocyte G0/G1 switch gene RP1-28O10.2; G0/G1 switch protein 2; G0/G1 switch regulatory protein 2; G0/G1switch 2; G0S2-like protein; Putative lymphocyte G0/G1 switch gene; putative lymphocyte G0/G1 switch protein 2
Gene Aliases: AI255151; AV006465; G0S2
UniProt ID: (Human) P27469, (Mouse) Q61585
Entrez Gene ID: (Human) 50486, (Mouse) 14373
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.