Novus Biologicals
Manufacturer Code:NBP2297680.05MG
Catalog # NB019458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7H7 |
Immunogen | Peptide corresponding to residues 110-123 of GiAlpha 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; Adenylate cyclase-inhibiting G alpha protein Gi Gi1 protein alpha subunit guanine nucleotide binding protein (G protein) alpha inhibiting activitypolypeptide 1 guanine nucleotide-binding protein G(i) subunit alpha-1; Gi1 protein alpha subunit; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; Guanine nucleotide-binding protein G(i) subunit alpha-1
Gene Aliases: Gi; GNAI1
UniProt ID: (Human) P63096
Entrez Gene ID: (Human) 2770
Molecular Function:
G-protein
enzyme modulator
heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.