Novus Biologicals
Manufacturer Code:NBP131601
Catalog # NBP131601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human G protein alpha Inhibitor 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; Adenylate cyclase-inhibiting G alpha protein Gi Gi1 protein alpha subunit guanine nucleotide binding protein (G protein) alpha inhibiting activitypolypeptide 1 guanine nucleotide-binding protein G(i) subunit alpha-1; Gi1 protein alpha subunit; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; Guanine nucleotide-binding protein G(i) subunit alpha-1
Gene Aliases: Gi; GNAI1
UniProt ID: (Human) P63096
Entrez Gene ID: (Human) 2770
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.