Novus Biologicals
Manufacturer Code:NBP270836G
Catalog # NBP270836G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6B3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-256 of human G protein alpha 16 (NP_002059) produced in E.coli. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Epididymis tissue protein Li 17E; G alpha-15; G alpha-15 G alpha-16 GNA16G-protein subunit alpha-15 G-protein subunit alpha-16 guanine nucleotide binding protein (G protein) alpha 15 (Gq class) guanine nucleotide-binding protein subunit alpha-15 Guanine nucleotide-binding protein subunit alpha-16; G alpha-16; G-protein subunit alpha-15; G-protein subunit alpha-16; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); Guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16
Gene Aliases: GNA15; GNA16
UniProt ID: (Human) P30679
Entrez Gene ID: (Human) 2769
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.